Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 2 x 45 mm, 26 Gauge
  100498-088
 :  62434-02
Inoculating Loops, Electron Microscopy Sciences
  100498-088
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 2 x 45 mm, 26 Gauge
$457.85
Each
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
  • Désignation:
    Inoculating Loops
  • Taille:
    2 x 45 mm
  • Jauge:
    26 Gauge