Diamond Points, Electron Microscopy Sciences
Diamond Points, Large, 2.9 mm, Round
  100497-978
 :  62155-29
Diamond Points, Electron Microscopy Sciences
  100497-978
 :  
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Large, 2.9 mm, Round
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
  • Ø:
    2.9 mm
  • Désignation:
    Diamond Points, Large
  • Shape:
    Round