Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences | Avantor
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
1/10 Hp Flex Shaft Machine Set, 110 V
Catalog # 100497-884
Fournisseur #:  62131-10
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Catalog # 100497-884
Supplier:  
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
1/10 Hp Flex Shaft Machine Set, 110 V
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A versatile hang-up flexible shaft machine.
 : Mounted accessories are on 2.35 mm (3/32") arbors.
 : Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
Spécifications
  • Désignation:
    1/10 Hp Flex Shaft Machine Set
  • Tension:
    110 V
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Standard Foot Speed Control
    Standard Foot Speed Control
    100497-914
    Log in pour la disponibilité
    Each - $199.37
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Probe Handle
    Probe Handle
    100497-882
    Log in pour la disponibilité
    Each - $47.20
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Flex Shaft
    Replacement Flex Shaft
    100497-888
    Log in pour la disponibilité
    Each - $122.59