Filing/Polishing Stick and Strips, Electron Microscopy Sciences | Avantor
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Filing and Polishing Stick, with two Strips (grit 120 and 240)
Catalog # 100497-846
Fournisseur #:  62118-02
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Catalog # 100497-846
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Filing and Polishing Stick, with two Strips (grit 120 and 240)
Price
91,67 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A plastic handle, 267 mm (10 1/2") long, holds strips of emery paper or cloth 279 x 25.4 mm (11 x 1"). Strips are easily replaced. Comes complete with 2 strips/set (grit 120 and 240).
Spécifications
  • Désignation:
    Filing and Polishing Stick, with two Strips (grit 120 and 240)