Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Folder
  100496-334
Pop-Up Slide Holder, Electron Microscopy Sciences
  100496-334
Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Holder, Electron Microscopy Sciences
Pop-Up Slide Folder
$323.02
Each
The Pop-Up holder holds 20 or 30 of 1 x 3" microscope slides in a numbered compartment. The compartments are recessed so the specimen cannot be touched when the cover is closed. Measurements: 8W x 131/2L x 11/32H".
  • Désignation:
    Pop-Up Slide Folder