Sample Storage Sets; Snap Cap Shell Vial, Electron Microscopy Sciences
Product Details & Documents
Poly-plug, Knurled snap-caps are friction fitted with glass shell vial, straight sided, and a flat bottom.
Alpha-numerical indexing system allows quick identification and location of stored items.
Ordering information: For vial sizes of 10 and 12 dram please call us for prices and availability.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Spécifications
- Avec bouchon:Oui
- Cap Type:Snap cap
- Capacité:10.40 ml
- Désignation:Shell vial series, dram 3