Perfect Scale, Electron Microscopy Sciences | Avantor
Perfect Scale, Electron Microscopy Sciences
Handle only, Perfect
Catalog # 100492-998
Fournisseur #:  70616-HD
Perfect Scale, Electron Microscopy Sciences
Catalog # 100492-998
Perfect Scale, Electron Microscopy Sciences
Perfect Scale, Electron Microscopy Sciences
Handle only, Perfect
Price
$333.62
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Perfect scale is made from Nickel alloy and 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD).
 : The Perfect Scale comes complete with one scale and a handle.
Spécifications
  • Désignation:
    Handle only, Perfect