Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Product Details & Documents
The Harris Uni-Core™ is a disposable multi-purpose sampling tool for soft substrates. Made from polypropylene, including a hollow body with built-in tool steel razor sharp cutting tip, with a plunger for the expelling of the punched sample.
- Individually wrapped
- Sterilized with ethylene oxide
- Reuse by rinsing the tip with ethanol and autoclaving for 20 minutes at 250 °F/15 psi
Spécifications
- Désignation:Sampling Tool 1.0