Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Desiccator Cabinet
Catalog # 100491-760
Supplier #:  71221
Low Temperature Dessicator, Electron Microscopy Sciences
Catalog # 100491-760
Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Dessicator, Electron Microscopy Sciences
Low Temperature Desiccator Cabinet
Price
$2,681.70
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm (1/4") acrylic with full-length hinges and stainless steel twist locking latches.
  • Easy viewing clear units or dark bonze for light sensitive materials
  • Deep, slide-in trays with perforated bottom
  • Available in three sizes
Specifications
  • Désignation:
    Low Temperature Desiccator Cabinet