Sample Bottles, PVC Transparent, Electron Microscopy Sciences
100 ml (3.4 oz.), 34 mm
  100490-548
 :  64264-10
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
  100490-548
 :  
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
Sample Bottles, PVC Transparent, Electron Microscopy Sciences
100 ml (3.4 oz.), 34 mm
These lightweight transparent PVC sample bottles come with snap-on tight-fitting caps and they offer excellent clarity, durability, and good chemical resistance. Maximum temperature –20 to 80 °C (–4 to 176 °F).
Note: The following sizes are also available: 30, 40, 48, 60, 90, 120,and 180 ml.
  • Capacité:
    100 ml (3.4 oz.)
  • Hauteur:
    34 mm