Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
20 Dram (74 ml), 64 mm, 38 mm
  100490-538
Supplier #:  64248-40
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
  100490-538
 :  
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
Polystyrene Containers; Poly-Containers, Electron Microscopy Sciences
20 Dram (74 ml), 64 mm, 38 mm
Lightweight, air-tight containers with snap-on polyethylene caps.
They have less resistance to breakage than polyethylene but they are transparent and ideal when used as a specimen container, storage vessel, or a block holder.
  • Ø int.:
    38 mm
  • Capacité:
    20 Dram (74 ml)
  • Hauteur:
    64 mm