Stabilizing Rings, Electron Microscopy Sciences | Avantor
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, Assorted Sizes
Catalog # 100489-790
Fournisseur #:  64440
Stabilizing Rings, Electron Microscopy Sciences
Catalog # 100489-790
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, Assorted Sizes
Price
$420.75
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
Spécifications
  • Désignation:
    Stabilizing Ring, Assorted Sizes