Tungsten Wire Filaments, Electron Microscopy Sciences
Tungsten coiled filaments, triple strand 6 turns, 6.3 mm (0.25"), 4"
  100489-588
 :  73938-3
Tungsten Wire Filaments, Electron Microscopy Sciences
  100489-588
 :  
Tungsten Wire Filaments, Electron Microscopy Sciences
Tungsten coiled filaments, triple strand 6 turns, 6.3 mm (0.25"), 4"
V-shaped filaments are high purity, vacuum grade tungsten wire 0.02" (0.50 mm) diameter, V-shaped filaments ½" (12.7mm) deep, with a bottom angle of 45°, either single wire or triple strand.
Coiled filaments are single or 3-strand coiled tungsten filaments, 0.5 mm (0.002") diameter wires.
  • Ø:
    6.3 mm (0.25")
  • Désignation:
    Tungsten coiled filaments, triple strand 6 turns
  • Longueur:
    4"