V-Vials®; Clear With Screw Cap, Electron Microscopy Sciences
Product Details & Documents
V-Vials are ideal for: micro mixing, homogenization, centrifugation, concentration, extraction, derivatization, radiosynthesis, hybridization, anaerobic reactions, and catalytic hydrogenation.
- All components can be autoclaved
- Graduated and clear
These vials are made from Wheaton-33 low extractable borosilicate glass and they safeguard against any change of product pH caused by alkali, which can be extracted from ordinary glass. Wheaton V-Vials are exceptionally sturdy vials with a conical interior bottom. Their special features insure maximum security and economy in handling, storing and shipping of costly micro and semi-micro samples.
Screw cap has solid top and TFE-faced has white (styrene butadiene) rubber liner.
Packaging: They come packed 12 vials in a hinged lid storage box.
Spécifications
- Avec bouchon:Oui
- Capacité:3.00 ml