Syringes Disposable, Electron Microscopy Sciences | Avantor
Syringes Disposable, Electron Microscopy Sciences
30 ml, Luerslip Eccentric
Catalog # 100489-468
Fournisseur #:  72513-LS
Syringes Disposable, Electron Microscopy Sciences
Catalog # 100489-468
Syringes Disposable, Electron Microscopy Sciences
Syringes Disposable, Electron Microscopy Sciences
30 ml, Luerslip Eccentric
Price
$175.90
Log in pour la disponibilité
Each Pack of 60
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These high quality disposable rubber piston syringes offer substantial savings.
  • Lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out
  • Syringes are ETO gas sterilized, latex and pyrogen free
  • Bold graduations are easily read and resist wipe-off
Packaging: All syringes are packaged in individual sterile strip packs.
Spécifications
  • Capacité:
    30 ml
  • Tip Style:
    Luerslip Eccentric