SB50 Grid Storage Box, Electron Microscopy Sciences
SB50 Grid Storage Box, Electron Microscopy Sciences
SB50 Grid Box with Unique Number
Catalog # 100489-236
Supplier #:  71136-12
SB50 Grid Storage Box, Electron Microscopy Sciences
Catalog # 100489-236
SB50 Grid Storage Box, Electron Microscopy Sciences
SB50 Grid Storage Box, Electron Microscopy Sciences
SB50 Grid Box with Unique Number
Price
$750.02
Log in for availability
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The clear cover can be rotated smoothly through 360 degrees once the slight initial resistance of the park position has been overcome exposing a maximum of 2 or 3 diamond shaped slots at any one time
The 50 diamond shaped slots have an alphanumeric referencing system. Each box is supplied with an index record card for additional information. The storage boxes are designed to be stacked, the base locating precisely over the face of another box. The material the bases are made from have been chosen due to their anti-static properties. The clear cover has self-lubricating properties.
Specifications
  • Désignation:
    SB50 Grid Box with Unique Number