Titer-Tops, Electron Microscopy Sciences
Titer-Tops, Electron Microscopy Sciences
Titer-Tops, 100, 112 µm, 6×3.25"
  100489-188
Supplier #:  70538-10
Titer-Tops, Electron Microscopy Sciences
  100489-188
Titer-Tops, Electron Microscopy Sciences
Titer-Tops, Electron Microscopy Sciences
Titer-Tops, 100, 112 µm, 6×3.25"
$232.93
Each Pack of 100
Titer-Tops prevents sample evaporation, well to well contamination, external contamination and sample spillage. Titer-Tops are made from 4.4 mil polyethylene film with an FDA approved non cytotoxic acrylic adhesive.
  • Titer-Tops are chemically resistant and thermally stable
  • Titer-Tops are gamma sterile
  • They are easily pierced with a micropipette tip to provide access to sample wells
Each pre-cut rectangular sheet is 6 x 3.25" with a paper liner. The center section of the liner measures 5 x 3.25" and once it is peeled off it leaves two hand-grip extension tabs on the two ends. These tabs allow for easy application of the Titer-Top to the plate. The acrylic adhesive is waterproof, solvent resistant, and compatible with all plastic microtiter plates. Titer-Tops will stay in place even when exposed to 100% humidity and temperatures ranging from -40 to 100 °C.
  • Désignation:
    Titer-Tops, 100
  • L×l:
    6×3.25"
  • Épaisseur:
    112 µm