4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Fluo-Green
  101413-206
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
  101413-206
 :  
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Fluo-Green
Biopsy processing cassettes with four square compartments each measuring 13 mm.
  • Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
  • Color:
    Fluo-Green