Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser, 120 ml Ea
  101412-922
 :  64299-01
Fluid Dispenser Bottles, Electron Microscopy Sciences
  101412-922
Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser Bottles, Electron Microscopy Sciences
Fluid Dispenser, 120 ml Ea
$85.82
Each
These ESD safe polyethylene solvent dispensers are available in three standard sizes.
  • Surface resistivity of 10E7 to 10E9
These dispensers have neither a stopper nor a cap to remove. Just touch the dispensing dish with a brush cloth or applicator and solvent is brought to the surface instantly as needed. Stainless steel valves seal contents in bottle until used. The fluid pumps into the dish and it does not drip back into the bottle, keeping stored liquids uncontaminated.
  • Désignation:
    Fluid Dispenser, 120 ml Ea