Glass Microbeakers, Electron Microscopy Sciences
Product Details & Documents
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Spécifications
- Ø ext.:21 mm
- Capacité:2.5 ml
- Hauteur:13 mm