Benchtop Mat, Electron Microscopy Sciences | Avantor
Benchtop Mat, Electron Microscopy Sciences
Benchtop Mat
Catalog # 100502-686
Fournisseur #:  64445
Benchtop Mat, Electron Microscopy Sciences
Catalog # 100502-686
Benchtop Mat, Electron Microscopy Sciences
Benchtop Mat, Electron Microscopy Sciences
Benchtop Mat
Price
$258.09
Log in pour la disponibilité
Each Roll of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions.
  • Sheets can be rolled up and discarded after use
  • Ideal for daily use in EM labs
Benchtop mat comes in a roll 50.8 cm (20") wide and 15.2 m (50') long in a dispenser carton, or a cut size of 20×24" sheets.
Spécifications
  • Désignation:
    Benchtop Mat