Square and Grid Scales, Electron Microscopy Sciences
NE34 Graticule, 21 mm, Grid (net) 0.1 mm squares
  100499-196
 :  68018-21
Square and Grid Scales, Electron Microscopy Sciences
  100499-196
 :  
Square and Grid Scales, Electron Microscopy Sciences
Square and Grid Scales, Electron Microscopy Sciences
NE34 Graticule, 21 mm, Grid (net) 0.1 mm squares
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.
  • Désignation:
    NE34 Graticule
  • Taille:
    21 mm
  • Divisions:
    Grid (net) 0.1 mm squares