Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Large Top-Loading Dispensing Bins
Catalog # 100498-780
Supplier #:  64388-18
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Catalog # 100498-780
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Top-Loading Dispensing Bins, Electron Microscopy Sciences
Large Top-Loading Dispensing Bins
Price
$462.59
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bins are made from ¹/₄" thick PVC. They are chemical resistant and easy to clean.
  • Removable top for easy loading
  • Hinged clear acrylic door
  • Different sizes for your needs
Ideal for pipette tips, small vials, cassette lids, cassette base molds, cotton swabs, aluminum stubs, embedding capsules etc.
Spécifications
  • Désignation:
    Large Top-Loading Dispensing Bins