Diamond Points, Electron Microscopy Sciences
Diamond Points, Inverted Wide, 8.3 mm, Cone
Catalog # 100497-990
 :  62155-83
Diamond Points, Electron Microscopy Sciences
Catalog # 100497-990
Supplier:  
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Diamond Points, Inverted Wide, 8.3 mm, Cone
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Spécifications
  • Ø:
    8.3 mm
  • Désignation:
    Diamond Points, Inverted Wide
  • Shape:
    Cone