Static Control Adjustable Wrist Band, Electron Microscopy Sciences | Avantor
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Static Control Adjustable Wrist Band, 12' Coil Cord
Catalog # 100497-456
Fournisseur #:  60622-12
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Catalog # 100497-456
Supplier:  
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Static Control Adjustable Wrist Band, 12' Coil Cord
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This hook and loop wrist strap is designed for those whom need to be static free in the working area.
This hook and loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1 MΩ resistor.
Spécifications
  • Désignation:
    Static Control Adjustable Wrist Band, 12' Coil Cord