Non-Marking Heel Grounder, Electron Microscopy Sciences | Avantor
Non-Marking Heel Grounder, Electron Microscopy Sciences
Heel Grounder with 1 Mega Ohm Resistor Blue
Catalog # 100497-448
Fournisseur #:  60619-10
Non-Marking Heel Grounder, Electron Microscopy Sciences
Catalog # 100497-448
Supplier:  
Non-Marking Heel Grounder, Electron Microscopy Sciences
Non-Marking Heel Grounder, Electron Microscopy Sciences
Heel Grounder with 1 Mega Ohm Resistor Blue
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
E.Z. Close is your low cost logical choice heel grounder.
  • The Velcro® fastening system is adjust-able and comfortable
  • Charge decay: less than 0.01 sec.; 2.4×108 Ω
They are made from premium grade non-marking conductive rubber that is highly abrasion and wear resistant.
Certifications: Meets DOD-263A standards (USA).
Delivery information: Includes: 18" conductive tab and 1¹/₂", two-layer, rubber cup. Built in resistor.
Spécifications
  • Désignation:
    Heel Grounder with 1 Mega Ohm Resistor Blue