Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, 20 mm id
  100496-214
Stabilizing Rings, Electron Microscopy Sciences
  100496-214
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Rings, Electron Microscopy Sciences
Stabilizing Ring, 20 mm id
$99.97
Each
Vinyl coated lead rings which stabilize glassware that is immersed in water baths; reduces tilting and chemical spills.
  • Désignation:
    Stabilizing Ring, 20 mm id