Sample Storage Sets; Square Bottle, Electron Microscopy Sciences
Product Details & Documents
A large size storage set. It comes with square clear glass bottles (1 ounce capacity) and black screw caps with plastic linings.
Alpha-numerical indexing system allows quick identification and location of stored items.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Spécifications
- Avec bouchon:Oui
- Capacité:78.00 ml
- Désignation:Square Bottle Series 2