Scalpel Handles Disposable, Electron Microscopy Sciences | Avantor
Scalpel Handles Disposable, Electron Microscopy Sciences
Disposable Blade Knife Handle #6, 5³⁄₁₆", Size 20, 21, 22, 23 blades
Catalog # 100493-602
Fournisseur #:  72040-06
Scalpel Handles Disposable, Electron Microscopy Sciences
Catalog # 100493-602
Scalpel Handles Disposable, Electron Microscopy Sciences
Scalpel Handles Disposable, Electron Microscopy Sciences
Disposable Blade Knife Handle #6, 5³⁄₁₆", Size 20, 21, 22, 23 blades
Price
$149.26
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These strong, moulded plastic scalpel handles provide a secure palm-fitting grip for post-mortem work, trimming light plaster casts, and similar forceful cutting.
Handle size #6 and #8 Bard-Parker* brand.
Spécifications
  • Désignation:
    Disposable Blade Knife Handle #6
  • Longueur:
    5³⁄₁₆"
  • For:
    Size 20, 21, 22, 23 blades