Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Wax Pen 1
Catalog # 100491-996
Supplier #:  72678
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Catalog # 100491-996
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Wax Pen 1
Price
$83.92
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This pen helps to flatten and separate tissue sections and reduces compression in thin sections.
  • Powered by AA batteries
 : Available in two models: Wax Pen 1 is powered by one AA battery; Wax Pen 2 is powered by two AA batteries.
Specifications
  • Désignation:
    Wax Pen 1