Quick Release Pipette Filler, Electron Microscopy Sciences
Product Details & Documents
This pipetting device provides an easy, precise way of filling and releasing liquid.
- Sizes are color coded
- Fits standard or disposable pipettes and diposable serological tooled-mouthpiece pipettes as per ASTM Standard E714-80
By rotating the knurled thumb wheel the liquid is drawn up or dispensed. The soft elastic chuck has a threaded collar that secures the plastic or glass pipette to the device.
Specifications
- Volume:25 ml
- Color:Red