Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences | Avantor
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Soap Wash Bottle, 500 ml (16.9 oz.)
Catalog # 100503-310
Fournisseur #:  64161
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Catalog # 100503-310
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Soap Wash Bottle, 500 ml (16.9 oz.)
Price
$33.73
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Permanently labeled/color-coded wash bottles.
Pack of six - Variety includes: Acetone, Methanol, Water, Iso-propyl Alcohol, Ethanol and Toluene. Toluene Wash Bottle is for short term storage only.
These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz. (500 ml), low density polyethylene.
Spécifications
  • Capacité:
    500 ml (16.9 oz.)
  • Désignation:
    Soap Wash Bottle