Tissue Chopper, Electron Microscopy Sciences
Product Details & Documents
The TC-1 Tissue Chopper is designed to facilitate the cutting of 100 to 1000 micron thick brain tissue slices for use in recording experiments.
The chopper includes a guillotine arm with adjustments to assure blade alignment. The brain sample is moved in respect to the arm with a stage whose position can be read with 2 to 3 micron precision on a digital micrometre scale. The stage is made with a removable "PTFE" surface plate to facilitate cleaning.
Spécifications
- Désignation:Replacement PFTE Plate