Microsurgical Knife, Electron Microscopy Sciences | Avantor
Microsurgical Knife, Electron Microscopy Sciences
Micro Surgical Knife 45°
Catalog # 101413-490
Fournisseur #:  72047-45
Microsurgical Knife, Electron Microscopy Sciences
Catalog # 101413-490
Microsurgical Knife, Electron Microscopy Sciences
Microsurgical Knife, Electron Microscopy Sciences
Micro Surgical Knife 45°
Price
$154.54
Log in pour la disponibilité
Each Pack of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from tool steel wire. Sharpened to microsurgical quality.
The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Packaging: 6 blister packs to a box.
Spécifications
  • Désignation:
    Micro Surgical Knife 45°