CAPINSERT™ for Cryovial® Tubes, Electron Microscopy Sciences
Red
  100501-126
 :  61805-R
CAPINSERT™ for Cryovial® Tubes, Electron Microscopy Sciences
  100501-126
CAPINSERT™ for Cryovial® Tubes, Electron Microscopy Sciences
CAPINSERT™ for Cryovial® Tubes, Electron Microscopy Sciences
Red
$60.92
Bag of 500 Each
Made from polypropylene
Color-coded inserts fit precisely into the cap of the CRYOVIAL® above for color identification. CapInsert® are sold 500 per bag.
  • Color:
    Red