Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
Catalog # 100491-748
Supplier #:  71155
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Catalog # 100491-748
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
Price
$45.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
  • Made from a special plastic that minimizes static
  • Complete with grid recording card
Specifications
  • Capacité:
    100 Grids
  • Désignation:
    Grid Storage Box
  • Dimensions:
    8 L×5 W×7 T mm