Modular Block for Dri-Bath, Electron Microscopy Sciences
Product Details & Documents
Fit all our Modular Block Dri-Baths.
- Anodized coating resists the harshest environments
- Versatile
- Solid anodized aluminum blocks provide maximum heat retention
- All blocks include thermometer well for easily measuring block temperature
- All blocks are the same size: Blocks measure: 3" (7.6cm) W x 2" (5cm) H x 4" (10.1cm) D
Spécifications
- Désignation:Solid Aluminum Block