Tin Can - Specimen Tin Boxes, Electron Microscopy Sciences | Avantor
Tin Can - Specimen Tin Boxes, Electron Microscopy Sciences
Specimen Tin Boxes, 1 oz
Catalog # 101412-454
Fournisseur #:  61020-02
Tin Can - Specimen Tin Boxes, Electron Microscopy Sciences
Catalog # 101412-454
Tin Can - Specimen Tin Boxes, Electron Microscopy Sciences
Tin Can - Specimen Tin Boxes, Electron Microscopy Sciences
Specimen Tin Boxes, 1 oz
Price
$218.39
Log in pour la disponibilité
Each Pack of 72
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A round seamless tin with a friction fit lid. Low form, flat bottoms.
  • White label on lid
These boxes are ideal for specimen or small item storage.
Spécifications
  • Désignation:
    Specimen Tin Boxes, 1 oz