Fixatives, Prepared, Electron Microscopy Sciences | Avantor
Fixatives, Prepared, Electron Microscopy Sciences
2.5% Glutaraldehyde in 0.1 M Millonigs Sodium Phosphate Buffer (pH 7.2)
Catalog # 100503-958
Fournisseur #:  15970
Fixatives, Prepared, Electron Microscopy Sciences
Catalog # 100503-958
CAS Number:  
Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences
2.5% Glutaraldehyde in 0.1 M Millonigs Sodium Phosphate Buffer (pH 7.2)
CAS Number:  
Price
$116.09
Log in pour la disponibilité
Each (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.
 : Certificates of analysis are available.
Spécifications
Spécifications
  • Désignation:
    2.5% Glutaraldehyde in 0.1 M Millonigs Sodium Phosphate Buffer (pH 7.2)