Partitioned Plastic Boxes, Electron Microscopy Sciences | Avantor
Partitioned Plastic Boxes, Electron Microscopy Sciences
5 Compartments, 5-7/8×3-1/2×15⁄16" (OVerall); 1-1/16×3×1-1/8" (Compartments)
Catalog # 100503-272
Fournisseur #:  64340
Partitioned Plastic Boxes, Electron Microscopy Sciences
Catalog # 100503-272
Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
5 Compartments, 5-7/8×3-1/2×15⁄16" (OVerall); 1-1/16×3×1-1/8" (Compartments)
Price
$50.88
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Organize singular or groups of items in individual compartments.
  • Extra-thick wall construction permits containment of heavy objects, protects delicate items
Stored items stay separately and are readily visible at all times.
Spécifications
  • Désignation:
    5 Compartments
  • Dimensions:
    5-7/8×3-1/2×15⁄16" (OVerall); 1-1/16×3×1-1/8" (Compartments)