Tri-Pour Beaker Caps, Electron Microscopy Sciences
Product Details & Documents
The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
The caps are available for all Tri-Pour beakers.
Specifications
- Fits:800 ml (27 oz.) beaker