Scintillation Vial Rack, Electron Microscopy Sciences
Welded polypropylene rack holds 20 ml scintillation vials.
- Autoclavable
- Resists nitric and hydrochloric acids
The hole measures ¹¹/₁₆" diameter.
- Désignation:Scintillation vial rack, white
- No. of Places:50
- W×D×H:12¾×6¾×1⅜"