Benchtop Mat, Electron Microscopy Sciences
Bench Top Mat Cut Size
  100500-862
Benchtop Mat, Electron Microscopy Sciences
  100500-862
Benchtop Mat, Electron Microscopy Sciences
Benchtop Mat, Electron Microscopy Sciences
Bench Top Mat Cut Size
$916.02
Box of 100 Each
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions.
  • Sheets can be rolled up and discarded after use
  • Ideal for daily use in EM labs
Benchtop mat comes in a roll 50.8 cm (20") wide and 15.2 m (50') long in a dispenser carton, or a cut size of 20×24" sheets.
  • Désignation:
    Bench Top Mat Cut Size