Grid Storage Box, 50 Capacity, Electron Microscopy Sciences
Product Details & Documents
Storage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Spécifications
- Capacité:50 Grids
- Désignation:Grid Storage Box, Clear Cover