Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
36 Compartments, 10×7×1-13/16" (Overall);1½×1-1/16×1-9/16" (Compartments)
  100496-202
Supplier #:  64344
Partitioned Plastic Boxes, Electron Microscopy Sciences
  100496-202
Partitioned Plastic Boxes, Electron Microscopy Sciences
Partitioned Plastic Boxes, Electron Microscopy Sciences
36 Compartments, 10×7×1-13/16" (Overall);1½×1-1/16×1-9/16" (Compartments)
$113.93
Each
Organize singular or groups of items in individual compartments.
  • Extra-thick wall construction permits containment of heavy objects, protects delicate items
Stored items stay separately and are readily visible at all times.
  • Désignation:
    36 Compartments
  • Dimensions:
    10×7×1-13/16" (Overall);1½×1-1/16×1-9/16" (Compartments)