Slide Staining Dish for 20 Slides, Electron Microscopy Sciences
Product Details & Documents
Ideal for immersing slides in several solutions.
The removable glass slide rack has an open bottom which allows for rapid filling and draining. It holds 20 slides-75 x 25 mm (3 x 1"), 75 x 38 mm (3 x 1 1/2"), or 75 x 51 mm (3 x 2") in size. The staining dish measures L x W x H: 107 x 87 x 70 mm. Slide Rack measures: 102 x 80 x 24 mm.
Ordering information: The complete set consists of 1x Staining dish, 1x Cover, 1x Slide Rack, and Handle.
Spécifications
- Désignation:Glass Slide Rack Only