Cotton Tipped Applicator, Electron Microscopy Sciences | Avantor
Cotton Tipped Applicator, Electron Microscopy Sciences
Large Tip, Wooden Shaft, 16 cm (6")
Catalog # 100488-464
Fournisseur #:  72309
Cotton Tipped Applicator, Electron Microscopy Sciences
Catalog # 100488-464
Cotton Tipped Applicator, Electron Microscopy Sciences
Cotton Tipped Applicator, Electron Microscopy Sciences
Large Tip, Wooden Shaft, 16 cm (6")
Price
$40.90
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft.
The pointed head size is 3/16" tapering to 1/32 x 5/8". Regular head size 3/16 x 5/8".
Packaging: Packed 100 sticks in a sealed bag.
Spécifications
  • Désignation:
    Large Tip, Wooden Shaft
  • Handle Length:
    16 cm (6")