Serum Bottles, Electron Microscopy Sciences
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
: Meets USP Type I standards.
- Capacité:100 ml
- Color:Amber
- Désignation:Serum Bottles