Fixatives, Prepared, Electron Microscopy Sciences
Formaldehyde/Glutaraldehyde, 2.5% each in 0.1 M Sodium Cacodylate Buffer (pH 7.4)
  100504-860
Fixatives, Prepared, Electron Microscopy Sciences
  100504-860
 :  
Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences
Formaldehyde/Glutaraldehyde, 2.5% each in 0.1 M Sodium Cacodylate Buffer (pH 7.4)
 :  
$135.87
Each (10ml)
Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.
 : Certificates of analysis are available.
  • Désignation:
    Formaldehyde/Glutaraldehyde, 2.5% each in 0.1 M Sodium Cacodylate Buffer (pH 7.4)