Brushes, Red Sable, Electron Microscopy Sciences
Product Details & Documents
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Spécifications
- Bristle Material:Sable hair
- Désignation:Red Sable Brush
- Taille:#5/0